0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Standards
Umweltanalyse-Standards
Diesel - Clay Loam 1, (CRM), Supelco®
Diesel - Clay Loam 1, (CRM), Supelco®
Diesel - Clay Loam 1, (CRM), Supelco®
Catalog # CRM558-100G
Supplier:
Merck
CAS Number:
Expand image
Diesel - Clay Loam 1, (CRM), Supelco®
Catalog # CRM558-100G
Supplier:
Merck
CAS Number:
Specifications
VE:
100 g
Bezeichnung:
Diesel - Clay Loam 1
Pk Info:
Pkg of 100 g
Typ:
Certified reference material
Specifications
About this item
Product Details
Product Details