ContactUsLinkComponent
BCECF (2′,7′ –bis-(Carboxyethyl)-5-(and-6)-carboxyfluorescein) ≥95% (durch HPLC) fluoreszierender pH-Indikator
Catalog # 51010.
Supplier:  Biotium
CAS Number:  
undefined
BCECF (2′,7′ –bis-(Carboxyethyl)-5-(and-6)-carboxyfluorescein) ≥95% (durch HPLC) fluoreszierender pH-Indikator
Catalog # 51010.
Supplier:  Biotium
CAS Number:  
Specifications
  • VE:
    1 mg
  • Seq:
    2070
Specifications
About this item
 :
 :520.44 g/mol
Storage Temperature:Gefrierschrank
CAS Number:85138-49-4
Formula:C₂₇H₂₀O₁₁
MDL Number:MFCD00036970