Order Entry
Austria
ContactUsLinkComponent
 
Supelco® Drug and metabolite standards, Dehydroepiandrosterone 3-sulfate (DHEAS) sodium salt solution, (CRM)
  D-065-1ML
 :  Merck
 :  
undefined
Supelco® Drug and metabolite standards, Dehydroepiandrosterone 3-sulfate (DHEAS) sodium salt solution, (CRM)
  D-065-1ML
 :  Merck
 :  

 

  • Konzentration:
    1,0 mg/ml in methanol (as free sulfate)
  • VE:
    1 mL
  • Normen:
    Certified reference material
  • CAS Nr.:
    78590-17-7
  • Bezeichnung:
    Dehydroepiandrosterone 3-sulfate (DHEAS) sodium salt solution
  • Bezeichnung:
    Dehydroepiandrosterone 3-sulfate (DHEAS) sodium salt solution
  • Pk Info:
    ampoule of 1 ml