LC columns, AdvanceBio RP-mAb, SB-C8
Supplier: Agilent
Certificates
About this item
AdvanceBio RP-mAb columns are designed to optimise the performance of intact and reduced mAb analysis when analysing monoclonal antibodies for biopharma discovery, development, and QA/QC applications.
- Superficially porous particles (3,5 µm) with wide pores (450 Å) increase mAb resolution while maintaining compatibility with all LC instruments
- Shorter analysis times compared to columns packed with fully porous particles of the same size
- Robust poroshell packed bed and 2 µm inlet frit extend column lifetime by helping prevent inlet blockage
The wide-pore poroshell technology of the column delivers high efficiency, a short analysis time, and low pressure, at temperatures below 80 °C.
Ordering information: Available in a range of chemistries, SB-C8, C4, and diphenyl.
Specifications
- Catalog No:
- HEWL787775-906
- HEWL785975-906
- HEWL785775-906
- HEWL783975-906
- HEWL783775-906
- HEWL789975-906
- HEWL789775-906
- Max. Druck:
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- pH-Bereich:
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- Temperaturlimit:
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- USP-Code:
- L7
- L7
- L7
- L7
- L7
- L7
- L7