Media in bottles, sabouraud dextrose broth
Artikelnr. LIOF402040
Lieferant: LIOFILCHEM
CAS-Nummer:
Spezifikationen
- VE:1
- Bezeichnung:Sabouraud dextrose broth, 6×100 ml
- Environmentally Preferable:
- Füllung:100 ml
Spezifikationen
Über diesen Artikel
Sabouraud dextrose broth (SDB) is a liquid medium used in qualitative procedures for isolation of yeasts and moulds and for the culture or subculture of fungi from clinical and nonclinical specimens.
Certifications: This medium conforms to the requirements of the harmonised method in the United States Pharmacopoeia (USP), European Pharmacopoeia (Ph. Eur.) and Japanese Pharmacopoeia (JP) for the microbiological examination of non sterile products.