Forceps, ceramic
145 mm, 1,5 mm
Artikelnr. 232-1644
Anbieter: SLG Kunststoff
Springe zu:
Product Details & Documents
Straight ceramic forceps with serrated tips.
- Anti-magnetic, corrosion-free, metal-free
- Dishwasher-safe
- Chemically resistant to aggressive media
- Extremely heat-resistant
- Non-wetting
Spezifikationen
- Breite:1,5 mm
- Länge:145 mm