Vial locator for GC autosampler
Artikelnr. PERCN6101182
Lieferant: PerkinElmer
Spezifikationen
- Bezeichnung:Vial locator for GC autosampler
- VE:1
Spezifikationen
Über diesen Artikel
This is a replacement vial locator for PerkinElmer AutoSystem GC autosamplers.
Included in the GC/MS PSS Injector Starter Kit (N6100447), the GC/MS CAP Injector Starter Kit for Clarus 680/580/480 (N6100448), and the GC/MS 590/690 Capillary Injector Consumable Kit (N6550448).
Compatible with:
Clarus 600
Clarus 400
Clarus 500
Autosystem
Autosystem XL
Clarus 680
Clarus 480
Clarus 590
Clarus 580
Clarus 690