EPA vials with PP cap
Catalog # SCERSP60961C-4
Supplier: DWK Life Sciences
Specifications
- Mit Deckel:Ja
- Verschlussmaterial:PP
- Verschlussgröße:24-400
- Ø×H:28×60 mm
- Septum:PTFE-faced silicone rubber
- Kapazität:20,00 ml
- Farbe:Klar
- Bezeichnung:EPA water analysis vials
- Verschlusstyp:Screw cap, open-top
- Verschluss-Farbe:Weiß
- VE:72
Specifications
About this item
These clear screw thread EPA vials are used for the collection and storage of water samples.
- Not pre-cleaned
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Certifications: These vials comply with the guidelines for establishing test procedures for the analysis of pollutants (Ref. EPA Methods 601, 602, 603 and 604).
Packaging: Vials are packaged in a plastic shrink wrap module, lehr-clean, as they come from the end of the production lehr and are placed into a corrugated shelf-pack.