EXP direct connect holder
Artikelnr. REST25808
Lieferant: Restek
Spezifikationen
- Bezeichnung:EXP direct connect holder
- VE:1 KIT
Spezifikationen
Über diesen Artikel
Holder for EXP guard cartridges.
- Free-Turn architecture lets you change cartridges by hand without breaking inlet/outlet fluid connections - no tools needed
- Patented titanium hybrid ferrules can be installed repeatedly without compromising high-pressure seal
- Auto-adjusting design provides ZDV (zero dead volume) connection to any 10-32 female port
Maximum holder pressure: 20000 psi (1400 bar).
Delivery: Includes hex-head fitting and two ferrules.