Order Entry
Austria
ContactUsLinkComponent
Drinking water odor standard (2 components)
Drinking water odor standard (2 components)
Catalog # REST30608
Supplier:  Restek
CAS Number:  
undefined
Drinking water odor standard (2 components)
Catalog # REST30608
Supplier:  Restek
CAS Number:  

Specifications

  • Konzentration:
    100 μg/ml in P and T methanol
  • VE:
    1 Ampoul
  • Bezeichnung:
    Drinking water odor standard
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

The standard consists of (±)-geosmin and 2-methylisoborneol (MIB).

  • Reference mix of the two most common odor-causing compounds
  • Convenient concentration for purge-and-trap analysis - 100 μg/ml in methanol
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.