Method 535 surrogate standard (dimethachlor ESA sodium salt)
Catalog # REST33203
Supplier: Restek
CAS Number:
Specifications
- Konzentration:100 μg/ml in methanol
- VE:1 Ampoul
- Bezeichnung:Dimethachlor ESA sodium salt
- Pk Info:1 ml/ampoule
- Typ:EPA method 535