NPI-1000 column test mix (8 components)
Artikelnr. REST35224
Lieferant: Restek
CAS-Nummer:
Spezifikationen
- VE:1
- Bezeichnung:NPI-1000 column test mix, 1000 µg/ml each in methylene chloride
- Pk Info:1 ml/ampoule
Spezifikationen
Über diesen Artikel
The mix consists of (C9:0) methyl nonanoate, (C13) n-tridecane, (C15) n-pentadecane, acenaphthylene, 4-chlorophenol, 1,6-hexanediol, 4-propylaniline and 1-undecanol.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.