WA EPH aliphatic hydrocarbon mix (6 components)
Catalog # REST31489
Supplier: Restek
CAS Number:
Specifications
- Konzentration:1000 μg/ml each in hexane
- VE:1
- Bezeichnung:WA EPH aliphatic hydrocarbon mix
- Pk Info:1 ml/ampoule
Specifications
About this item
The mix consists of, (C8) n-octane, (C10) n-decane, (C12) n-dodecane, (C16) n-hexadecane, (C21) n-heneicosane, (C34) n-tetratriacontane.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.