ContactUsLinkComponent
Calibration mix D3710-95 (16 components)
Artikelnr. REST31223
Lieferant:  Restek
CAS-Nummer:  
undefined
Calibration mix D3710-95 (16 components)
Artikelnr. REST31223
Lieferant:  Restek
CAS-Nummer:  
Spezifikationen
  • VE:
    1
  • Bezeichnung:
    D3710-95 calibration mix
  • Pk Info:
    1 ml/ampoule
Spezifikationen
Über diesen Artikel
These calibration mixtures are made with pure, highly characterized neat material and are prepared using NIST-traceable balances and weights. Each ampul is supplied with a data sheet indicating the exact concentration and a sample chromatogram.
D3710-95 calibration mix contains (C5) n-pentane (109-66-0), 8%; (C6) n-hexane (110-54-3), 6%; (C7) n-heptane (142-82-5), 10%; (C8) n-octane (111-65-9), 5%; (C10) n-decane (124-18-5), 4%; (C12) n-dodecane (112-40-3), 4%; (C13) n-tridecane (629-50-5), 2%; (C14) n-tetradecane (629-59-4), 2%; (C15) n-pentadecane (629-62-9), 2%; n-butylbenzene (104-51-8), 4%; 2,4-dimethylpentane (108-08-7), 6%; 2-methylbutane (isopentane) (78-78-4), 10%; 2-methylpentane (107-83-5), 6%; n-propylbenzene (103-65-1), 5%; toluene (108-88-3), 12%; p-xylene (106-42-3), 14%.
Certifications: ASTM Methods D2887 and D3710-95. Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.