EPA 539 calibration stock standard (7 components)
REST31998
: Restek
:
- VE:1 Ampoul
- Bezeichnung:EPA 539 calibration stock standard
- Pk Info:1 ml/ampoule
Contains the seven hormones listed in EPA Method 539 at the appropriate concentrations. The seven components are 4-androstene-3, 17-dione, equilin, 17-beta-Estradiol, estriol, estrone, 17-alpha-ethynylestradiol and testosterone.
: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
: UCMR3 method 539 calibration standard with concentrations of 10 to 400 µg/ml to more conveniently match UCMR3 requirements (cat.# 32461) also available.