QuEChERS internal standard mix for GC/NPD and LC/MS/MS analysis (2 components)
Catalog # REST33266
Supplier: Restek
CAS Number:
Specifications
- Konzentration:In acetonitrile
- VE:1 Ampoul
- Bezeichnung:QuEChERS internal standard mixture for GC/NPD and LC/MS/MS analysis
- Pk Info:5 ml/ampoule
Specifications
About this item
Ready to use for QuEChERS extractions—no dilutions necessary.
It consists of triphenylphosphate (20 μg/ml) and tris-(1,3-dichloroisopropyl)phosphate (50 μg/ml).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.