Express Eingabe
Austria
AuftragsverfolgungKontakt
Pharmaceutical mix #2 (4 components)

Pharmaceutical mix #2 (4 components)

Pharmaceuticals mix #2, 200 µg/ml each in P and T methanol, 1 ml/ampoule, 1 mL
Catalog # REST31118
Supplier:   Restek
Pharmaceutical mix #2 (4 components)
Catalog # REST31118
Supplier:  Restek
CAS Number:  
Pharmaceutical mix #2 (4 components)
Pharmaceutical mix #2 (4 components)

Pharmaceuticals mix #2, 200 µg/ml each in P and T methanol, 1 ml/ampoule, 1 mL

CAS Number:  
Price
48,50 €
Log in for availability
1 * 1 mL
Product Details & Documents

This pharmaceutical mix #2 contains four components: Gemfibrozil, Ibuprofen, Naproxen and Triclosan.

Shelf life maximum 24 months and minimum 6 months.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.

Specifications

Specifications

  • Konzentration:
    200 µg/ml each in P and T methanol
  • VE:
    1 mL
  • Bezeichnung:
    Pharmaceuticals mix #2
  • Pk Info:
    1 ml/ampoule

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.