Order Entry
Austria
ContactUsLinkComponent
 
Pesticide mix #2 (rev), organochlorine, method 525.2 (8 components)
Catalog # REST33011
Supplier:  Restek
CAS Number:  
undefined
Pesticide mix #2 (rev), organochlorine, method 525.2 (8 components)
Catalog # REST33011
Supplier:  Restek
CAS Number:  

Specifications

  • Konzentration:
    500 µg/ml each in acetone
  • VE:
    1 Ampoul
  • Bezeichnung:
    Pesticide mix #2, organochlorine
  • Pk Info:
    1 ml/ampoule
  • Typ:
    Pesticide mixture

Specifications

About this item

Contains: Chlorobenzilate, chloroneb, chlorothalonil, DCPA methyl ester (chlorthal-dimethyl), heptachlor epoxide (isomer A), trans-nonachlor, cis-permethrin, trans-permethrin.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.