Iodomethane
Iodomethane, 2000 μg/ml in P and T methanol, 1 ml/ampoule, 1 mL
Artikelnr. REST30292
Anbieter: Restek
Springe zu:
Product Details & Documents
Zertifizierung: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Spezifikationen
Spezifikationen
- Konzentration:2000 μg/ml in P and T methanol
- VE:1 mL
- Bezeichnung:Iodomethane
- Pk Info:1 ml/ampoule
- Cat. No.:REST30292
- CAS Nr.:74-88-4
- Formel:CH₃I
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



