ASTM D3606 calibration standard #2 with sec-butanol internal standard (3 components)
REST30662
: Restek
:
- VE:1 mL
- Bezeichnung:ASTM D3606 calibration standard #2 with sec-butanol internal standard
- Pk Info:1 ml/ampoule
Consists of three components, those are benzene, 2-butanol and toluene.
: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.