GC multiresidue pesticide kit
Catalog # REST32562
Supplier: Restek
CAS Number:
Specifications
- VE:1
- Bezeichnung:GC multiresidue pesticide kit
Specifications
About this item
Accurately identify and quantify pesticide residues by GC-MS/MS in fruits, vegetables, botanicals, and herbals such as tea, ginseng, ginger, echinacea, and dietary supplements.
- Formulated and grouped for maximum long-term stability
- Quantitatively tested to confirm composition
- Contains 1 ml each of these mixtures
GC multiresidue pesticide kit consist of GC multiresidue pesticide standard #1, GC multiresidue pesticide standard #2, GC multiresidue pesticide standard #3.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements. Comprehensive 203-compound kit covers food safety lists by the FDA, USDA.