Heptachlor
Heptachlor, 1000 µg/ml in methanol, 1 ml/ampoule, 1
REST32228
: Restek
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
- Konzentration:1000 µg/ml in methanol
- VE:1
- Bezeichnung:Heptachlor
- Pk Info:1 ml/ampoule
- Cat. No.:REST32228
- CAS Nr.:76-44-8
- Bezeichnung:Heptachlor
- Formel:C₁₀H₅Cl₇



