Express Eingabe
Austria
AuftragsverfolgungKontakt
Fortification Mix (7 components) | Avantor

Fortification Mix (7 components)

Fortification Mix, 2000 µg/ml each in methylene chloride, CRM, 1 ml/ampoule, 1 Ampoul
Artikelnr. REST31813
Anbieter:   Restek
Fortification Mix (7 components)
Artikelnr. REST31813
Anbieter:  Restek
CAS Nummer:  
Fortification Mix (7 components)
Fortification Mix (7 components)

Fortification Mix, 2000 µg/ml each in methylene chloride, CRM, 1 ml/ampoule, 1 Ampoul

CAS Nummer:  
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1 Ampull
Product Details & Documents

EPA Contract Lab Program (CLP) Statement of Work (SOW) OLC03.2 is an analytical method for the detection of low-concentration volatile, semivolatile, and pesticide/Aroclor organics in aqueous samples using deuterated monitoring compounds (DMCs) as sample-by-sample accuracy indicators. Restek supports this method with a convenient VOA MegaMix (with m- and p-xylene isomers at half the concentration of other components), a deuterated monitoring compounds (DMCs) mix, a fortification mix for all compounds requiring calibration at higher concentration, and single-compound solutions of
3,3'-dichlorobenzidine, and 4-chloroaniline.

This fortification mix contains: 4,6-Dinitro-2-methylphenol (Dinitro-o-cresol), 2,4-Dinitrophenol, 2-Nitroaniline, 3-Nitroaniline, 4-Nitroaniline, 4-Nitrophenol, 2,4,5-Trichlorophenol.

Spezifikationen

Spezifikationen

  • Konzentration:
    2000 µg/ml each in methylene chloride
  • VE:
    1 Ampoul
  • Normen:
    CRM
  • Bezeichnung:
    Fortification Mix
  • Pk Info:
    1 ml/ampoule

Spezifikationen

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.