PA DEP UST standard (11 components)
Catalog # REST30433
Supplier: Restek
CAS Number:
Specifications
- VE:1
- Bezeichnung:PA DEP UST standard (11 components), 2000 μg/ml in P and T methanol
- Pk Info:1 ml/ampoule
Specifications
About this item
Standard contains benzene, 1,2-dibromoethane (EDB), 1,2-dichloroethane, ethylbenzene (100-41-4), isopropyl benzene (cumene), methyl tert-butyl ether (MTBE), naphthalene, toluene, m-xylene, o-xylene, p-xylene.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.