GC multi-residue pesticide standard #8 (24 components)
Catalog # REST32570
Supplier: Restek
CAS Number:
Specifications
- VE:1 mL
- Bezeichnung:GC multiresidue pesticide standard #8, 24 components, 100 μg/ml each in toluene
- Pk Info:1 ml /ampoule
Specifications
About this item
The multiresidue accurately identify and quantify pesticide residues by GC-MS/MS in fruits, vegetables, botanicals, and herbals such as tea, ginseng, ginger, echinacea, and dietary supplements.
- Quantitatively tested to confirm composition, detailed support documentation provided
- Formulated and grouped for maximum long-term stability
The multiresidue pesticide standard consist of 24 components bromfenvinfos-methyl, bromfenvinphos, bromophos ethyl, bromophos methyl, carbophenothion, chlorfenvinphos, chlorthiophos, coumaphos, edifenphos, ethion, fenamiphos, fenchlorphos (ronnel), fenthion, iodofenphos, leptophos, malathion, methacrifos, profenofos, prothiofos, sulphotepp, sulphrofos, terbufos, tetrachlorvinfos, tolclofos-methyl.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.