Disinfectant, ClearKlens Tego 2000 SC, VH25
Artikelnr. 141-1447
Lieferant: Diversey
Spezifikationen
- Bezeichnung:ClearKlens Tego 2000 SC disinfectant, VH25
- VE:50 Bottle
- pH-Wert:8,0 (20 °C)
Spezifikationen
Über diesen Artikel
ClearKlens Tego 2000 SC is a concentrated disinfectant for the application in the pharmaceutical and medical industries.
- Sterile, amphoteric disinfectant
- Detex label for proof of irradiation
- Triple packed to facilitate transport to the clean room
- No discolouration, low toxicity and odourless
- Broad spectrum of efficacy
- Effective in hard and soft water
ClearKlen's Tego 2000 SC is 0,2 μm filtered and triple packaged, validated gamma irradiation process. It is suitable for spray and nebulisation applications.
Caution: Biozidprodukte vorsichtig verwenden. Vor Gebrauch stets Etikett und Produktinformationen lesen.