Alkaline detergents, powder, LaboClean PLM, neodisher®
148-3133
- Bezeichnung:LaboClean PLM, alkaline detergent
- Seq:02
- Packung:Bucket
- VE:10 kg
- pH-Bereich:11,1 - 11,6
Mildly alkaline detergent.
- Free of oxidative additives
Suitable for the removal of the basic components of creams respective lotions and easily emulsified residues. With defoaming properties.