ContactUsLinkComponent
LC OQ wavelength accuracy standard
  REST31053
 :  Restek
 :  
LC OQ wavelength accuracy standard
  REST31053
 :  Restek
 :  
  • VE:
    1 Ampoul
  • Bezeichnung:
    Erbium perchlorate (14017-55-1), 10 mg/ml in water
  • Pk Info:
    5 ml/ampoule
Erbium perchlorate, 14017-55-1