α-BHC, reference standard
α-BHC, 1000 µg/ml in methanol, 1 ml/ampoule, 1
REST32206
: Restek
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
- VE:1
- Bezeichnung:α-BHC, 1000 µg/ml in methanol
- Pk Info:1 ml/ampoule
- Cat. No.:REST32206
- CAS Nr.:319-84-6
- Bezeichnung:α-BHC
- Formel:C₆H₆Cl₆



