Natriumhydroxid Konzentrat zur Verdünnung zu 1 L, 0.1 M NaOH (0.1 N) volumetrische Lösung, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Boiling Pt:100 °C
Melting Pt:–10–14 °C
Density:1,07 g/cm³ (20 °C)
Storage Temperature:Raumtemperatur
UN:1824
ADR:8,II
:GHS05
Specification Test Results
- Factor:0.998 - 1.002
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Specification Test Results
Frequently Bought Together