Express Eingabe
Austria
AuftragsverfolgungKontakt
Appendix IX mix #1, revised

Appendix IX mix #1, revised

Appendix IX Mix #1, (17 components) 2000 µg/ml each in methylene chloride, EPA Appendix IX, 1 ml/ampoule, 1 mL
Catalog # REST32459
Supplier:   Restek
Appendix IX mix #1, revised
Catalog # REST32459
Supplier:  Restek
CAS Number:  
Appendix IX mix #1, revised

Appendix IX Mix #1, (17 components) 2000 µg/ml each in methylene chloride, EPA Appendix IX, 1 ml/ampoule, 1 mL

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 mL
Product Details & Documents

Appendix IX mix #1 includes 17 compounds.

Specifications

Specifications

  • VE:
    1 mL
  • Bezeichnung:
    Appendix IX Mix #1, (17 components) 2000 µg/ml each in methylene chloride
  • Pk Info:
    1 ml/ampoule
  • Typ:
    EPA Appendix IX

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.