Preparative HPLC columns, Ultra Biphenyl
Lieferant: Restek
Zertifikate
Über diesen Artikel
These biphenyl columns offer a greater degree of dispersion than conventional phenyls and a greater degree of polarizability than phenyl hexyls, creating higher selectivity and a greater range of usability. Because of these heightened interactions, this column shows substantial increases in retention—especially for dipolar, unsaturated, or conjugated solutes—and enhanced orthogonal selectivity when using methanol mobile phases.
- Easy scale-up from analytical columns
Ideal for increasing sensitivity and selectivity in LC-MS analyses.
Spezifikationen
- Catalog No:
- REST9109518
- REST9109519
- REST9109577
- Kohlenstoffbelastung:
- 15%
- 15%
- 15%
- Endcapped:
- Ja
- Ja
- Ja
- pH-Bereich:
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- Surface area:
- 300 m²/g
- 300 m²/g
- 300 m²/g
- Temperaturlimit:
- 80 °C
- 80 °C
- 80 °C
- USP-Code:
- L11
- L11
- L11