Hand disinfectant, MYXAL® SEPT 80
FAWE13661-010
- Bezeichnung:Hand disinfectant, biocide, MYXAL SEPT 80, 1000 ml bottle
- VE:1
This biocide is skin-friendly due to the combination of active ingredients in a position to achieve a broad spectrum of action with simultaneous skin care by moisturising substances.
- Used for hygienic hand disinfection
- Recipe is water-clear, perfume and phenol free
- Highly flammable liquid and vapour
It is bactericidal (including MRSA), tuberculocidal, yeasticidal, yeasticidal according to European standard and DVV / RKI guideline incl. Noro and Rota viruses and also listed at the Association for Applied Hygiene (VAH).
Can be stored in original sealed containers at room temperature. Polyethylene bottles can be disposed as polyethylene waste when empty.
Ingredients: 80 g ethanol, 0.1 g alkyl (C12-16) dimethylbenzylammonium chloride in 100 g solution
: Use MYXAL® SEPT 80 safely, always read the label and product information before use. Intended for external use only. Avoid eye contact. Keep out of the reach of children. Do not use MYXAL® SEPT 80 after expiry date date. Do not use in case of hypersensitivity to any of the ingredients.