AuftragsverfolgungKontakt
Capillary column installation kit | Avantor
Capillary column installation kit
Capillary column installation kit with diamond-tipped cutting pencil
Artikelnr. HEWL430-2000
Anbieter:   Agilent
Capillary column installation kit
Artikelnr. HEWL430-2000
Anbieter:  Agilent
Capillary column installation kit
Capillary column installation kit with diamond-tipped cutting pencil
Preis
290,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Lieferumfang: This kit includes a diamond-tipped pencil, 20x magnifier, ferrule removal tool, septum removal tool, refillable 60 ml bottle of snoop.
Spezifikationen
  • Bezeichnung:
    Capillary column installation kit with diamond-tipped cutting pencil