Organic reference standard, glycopyrronium bromide, Mikromol™
Catalog # PROOMM0834.00
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- VE:100 mg
- Normen:Reference standard
- CAS Nr.:51186-83-5
- Bezeichnung:Glycopyrronium bromide
- Formel:C₁₉H₂₈NO₃.Br
- Bezeichnung:Glycopyrronium bromide
- Molekulargewicht (Mw):398,33 g/mol
Specifications
About this item
Certifications: Accredited by ISO 9001.