Coated slides
Indium tin oxide coated slides, 25×25 mm
Artikelnr. SIAL703176-10PAK
Anbieter: Merck
Springe zu:
Product Details & Documents
Indium tin oxide coated glass slides.
- Good resistivity of 70-100 Ω/sq
- Unpolished surface with 86 % transmittance
Spezifikationen
- B×T:25×25 mm
- Bezeichnung:Indium tin oxide coated slides