Ringers solution ¹/₄ strength tablets, NutriSelect® Basic, Millipore®
Artikelnr. SIAL96724-100TAB
Lieferant: Merck
CAS-Nummer:
Spezifikationen
- VE:100 Tab.
- Bezeichnung:Ringers solution 1/4 strength tablets
Spezifikationen
Über diesen Artikel
Ringers solution maintains the osmotic balance for bacteria.
- One tablet is sufficient for 500 ml
Used for the preparation of suspension of samples and for use as a diluent in dilution techniques for enumeration. Also used in the microbial control of apparatus by the rinse and swab method.