Membrane dispenser for microbiological control, electronic butler (eButler), Whatman™
Catalog # WHAT10477103
Supplier: Whatman products (Cytiva)
Specifications
- Bezeichnung:Electronic butler (eButler) membrane dispenser
- VE:1
Specifications
About this item
The eButler rapid membrane dispenser allows for more efficient microbiological testing. It rapidly dispenses culture membranes using either hands-free or push-button operation.
- Aluminum and stainless steel construction
- Designed for easy membrane loading and device usability
- Up to 400 membranes per package
- Compatible with Whatman™ sterile mixed cellulose ester (MCE) microbiology membranes
This is the only eButler compatible with Whatman™ MCE microbiology membranes.