WiFi streamer, Dino-Lite WF-10
Catalog # 630-2923
Supplier: Dino-Lite / IDCP B.V.
Specifications
- Bezeichnung:Wi-Fi-Streamer mit Bedienungsanleitung, Netzteil und 1880 Ah-Li-Ion-Batterie
- Seq:901
- VE:1
Specifications
About this item
WF-10 allows compatible Dino-Lite USB microscopes to be used wirelessly in connection with an iOS (iPhone/iPad) or Android app on any tablet, smartphone, or even in the DinoCapture software for Windows computers.
- Can be used anywhere to create a WiFi signal for streaming the Dino-Lite live images
- Can stream from one Dino-Lite microscope to multiple devices
- Built-in battery is replaceable
Ideal for field work or presentations.
Compatible with: AM/AD4xxx (R4), AM4xx5, AM/AD7xxx (R4), MEDL4xx (R4), MEDL7xx (R4) series.
Please note: The free DinoConnect App needs to be downloaded from the App Store or Google Play.