5-Bromisatin
Marketsource
Anbieter: CHEMPUR
Springe zu:
Über diesen Artikel
Synonyme: 5-Bromoindoline-2,3-dione, 5-Bromo-1H-indole-2,3-dione
- CAS Nummer:87-48-9
- Formel:C₈H₄BrNO₂
- MDL-Nummer:MFCD00149345
- Molekulargewicht:226.03 g/mol
- EINECS:201-747-7
2 Optionen unten verfügbar
Product Details & Documents
Synonyme: 5-Bromoindoline-2,3-dione, 5-Bromo-1H-indole-2,3-dione
- CAS Nummer:87-48-9
- Formel:C₈H₄BrNO₂
- MDL-Nummer:MFCD00149345
- Molekulargewicht:226.03 g/mol
- EINECS:201-747-7
Product Family Options
Produktinformationen
- PackungVEVerfügbarkeitPreis
- Spezifikationen:MarketsourceMarketsource
This product has been blocked by your organization. Please contact your purchasing department for more information



