Order Entry
Austria
ContactUsLinkComponent
2-Methyl-1-Propanol, Sigma-Aldrich®
Flammable
Corrosive
Irritant
2-Methyl-1-Propanol, Sigma-Aldrich®
Catalog # SIALW217913-1KG-K
Supplier:  Merck
CAS Number:  
undefined
Flammable
Corrosive
Irritant
2-Methyl-1-Propanol, Sigma-Aldrich®
Catalog # SIALW217913-1KG-K
Supplier:  Merck
CAS Number:  

Specifications

  • Packung:
    Glasflasche für Feststoffe
  • VE:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :74.12 g/mol
Merck Index:13,05148
Boiling Pt:107,9 °C (1013 hPa)
Melting Pt:–108 °C
Density:0,802 g/cm³ (20 °C)
Flash Pt:28 °C
Storage Temperature:Raumtemperatur
EINECS:201-148-0
UN:1212
ADR:3,III
 :M-Clarity™ Qualitätsstufe = MQ300
 :GHS02
 :GHS05
 :GHS07
CAS Number:78-83-1
Formula:(CH₃)₂CHCH₂OH
MDL Number:MFCD00004740