2-Propylvaleriansäure
Glass bottle for solids, 1 mg
Artikelnr. FITZ10-V12-M605121
Anbieter: FITZGERALD INDUSTRIES INTL
2-Propylvaleriansäure
Artikelnr. FITZ10-V12-M605121
Anbieter: FITZGERALD INDUSTRIES INTL
CAS Nummer: 99-66-1
Springe zu:
Product Details & Documents
Synonyme: 2-Propylpentanoic acid, Valproic Acid, 2,2-Di-n-propylacetic acid
- CAS Nummer:99-66-1
- Formel:C₈H₁₆O₂
- MDL-Nummer:MFCD00002672
- Molekulargewicht:144.21 g/mol
- Merck-Index:12,10049
- Siedepunkt:220 °C (1013 hPa)
- Dichte:0,905 g/cm³ (20 °C)
- Blitzpunkt:111 °C
- Lagertemperatur:Raumtemperatur
- EINECS:202-777-3
- UN:3265
- ADR:8,III
Spezifikationen
Spezifikationen
- Packung:Glass bottle for solids
- VE:1 mg
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



