

N,N-Dimethylformamiddipropylacetal ≥97,5% zur GC-Derivatisierung, Supelco®
Catalog # 394998-10X1ML
Supplier: Merck
CAS Number:
Specifications
- Packung:Glasflasche
- VE:10 mL
- Seq:2275
Specifications
Specification Test Results
- APPEARANCE (COLOR):Colorless
- APPEARANCE (FORM):Liquid
- PURITY (GC AREA %):≥ 97.5 %
- INFRARED SPECTRUM:CONFORMS TO STRUCTURE
Specification Test Results
About this item
:
:175.27 g/mol
Boiling Pt:67…68 °C (14 mmHg) (Lit.)
Density:0,854 g/mL (25 °C) (Lit.)
ADR:3,III
:M-Clarity™ Qualitätsstufe = MQ100
EINECS:227-855-4
UN:1993
:GHS02
:GHS07
CAS Number:6006-65-1
Formula:C₉H₂₁NO₂
MDL Number:MFCD00009374