Aluminiumkaliumsulfat Dodecahydrat
Catalog # ROTHCN78.1
Supplier: Roth Carl
CAS Number:
Specifications
- Packung:Glass bottle for solids
- VE:500 g
- Seq:3645
Specifications
About this item
:
:474.39 g/mol
Merck Index:12,00373
Melting Pt:92.5 °C
Density:1.72 g/cm³ (20 °C)
EINECS:233-141-3
CAS Number:7784-24-9
Formula:AlK(SO₄)₂·12H₂O
MDL Number:MFCD00149143