Sigma-Aldrich® Select yeast extract, BioReagent, suitable for cell culture
Lieferant: Merck
Zertifikate
Über diesen Artikel
Select Yeast extract is used as a bacteriological media component for a variety of microorganisms and molecular genetics applications.
- Spray dried powder, autolyzed yeast extract
Water soluble portion of autolyzed yeast with intact B-complex vitamins. Yeast extract is a mixture of amino acids, peptides, water soluble vitamins and carbohydrates and can be used as additive for culture media.
Packaging: Packed in poly bottles.
Frequently Bought Together