m-Endo agar LES, NutriSelect® Plus, Millipore®
Catalog # SIAL85766-500G-F
Supplier: Merck
CAS Number:
Specifications
- VE:500 g
- Bezeichnung:m-Endo Agar LES
Specifications
About this item
m-Endo agar LES can be used for the enumeration of coliforms in water using the membrane filter technique for coliform enumeration.
- Selective and differential for Escherichia coli and Coliforms; Proteus and Salmonella
- Final pH: 7,2±0,2 at 25 °C
The membare composition is agar - 15 g/l; casein hydrolysate - 3,7 g/l; di-potassium hydrogen phosphate - 3,3 g/l; lactose - 9,4 g/l; pararosanilin (fuchsin) - 0,8 g/l; peptone from meat - 3,7 g/l; potassium hydrogen phosphate, 1 g/l; sodium chloride - 3,7 g/l; sodium lauryl sulfate - 0,05 g/l; sodium sulphite - 1,6 g/l; tryptose - 7,5 g/l and yeast extract - 1,2 g/l.