NapSul-Ile-Trp-CHO
ENZOBMLPI1250005
:
- VE:5 mg
- Seq:04
- Protein/Peptid-Name:NapSul-Ile-Trp-CHO
- Purity:98%
Cathepsin inhibitor
Potent and selective inhibitor of cathepsin L, a lysosomal cysteine protease. IC50=1,9 nM.
Cathepsin inhibitor
Potent and selective inhibitor of cathepsin L, a lysosomal cysteine protease. IC50=1,9 nM.