AuftragsverfolgungKontakt
8-Br-cGMP ≥96% (durch HPLC)
Artikelnr.
8-Br-cGMP ≥96% (durch HPLC)
Artikelnr.
Anbieter:  
CAS Nummer:  51116-01-9
8-Br-cGMP ≥96% (durch HPLC)
8-Br-cGMP ≥96% (durch HPLC)
CAS Nummer:  51116-01-9
Product Details & Documents
A cell-permeable cGMP analog which is more resistant to phosphodiesterases than cGMP and which preferentially activates cGMP-dependent protein kinase. It inhibits thrombin stimulated arachidonic acid release in human platelets.
  • PKG activator
White to off-white powder
Soluble in water (25mg/ml) or aqueous buffers. Sparingly soluble in methanol (4mM). Insoluble in DMSO, dimethyl formamide and 96% ethanol.
  • CAS Nummer:51116-01-9
  • Formel:C₁₀H₁₂BrN₅NaO₈P
  • Molekulargewicht:464.101 g/mol
  • Lagertemperatur:Gefrierschrank