Human recombinant Cathepsin L1
Catalog # ENZOENZPRT1690010
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- VE:10 µG
- Protein/Peptid-Typ:Rekombinant
- Spezies:Human
- Seq:02
- Proteinsynonym:MEP|CTSL1|CATL|CTSL
- Protein/Peptid-Name:Cathepsin L1
- Purity:>98% (SDS-PAGE)
- Molecular weight:~36,7 kDa
- Zusammensetzung:Lyophilized from 0,22 µm filtered solution in 50 mM sodium acetate, pH 5,0, containing 100 mM NaCl. Generally 5 - 8% mannitol or trehalose is added as a protectant before lyophilization.
Specifications
About this item
Produced in HEK 293 cells.