Native alphaC. difficilealpha toxin A from ribotype 027
Artikelnr. ENZOENZPRT1080002
Lieferant: ENZO LIFE SCIENCES
CAS-Nummer:
Spezifikationen
- VE:2 µG
- Protein/Peptid-Typ:Nativ
- Seq:02
- Proteinsynonym:Clostridium difficile toxin A|tcdA|toxA|native antigen
- Protein/Peptid-Name:alphaC. difficilealpha toxin A from ribotype 027
- Purity:>95% (SDS-PAGE)
- Zusammensetzung:Lyophilised, Contains 0,05 M HEPES, 0,15 M sodium chloride and 5% sucrose
- Reinigung:Purified protein
Spezifikationen
Über diesen Artikel
Highly purifed preparation from hypervirulent ribotype 027
- High potency
- Minimal lot to lot variation
- Single lot bulk quantites available
- Lypohilized for ease of handling and long term stability
From Clostridium difficile, strain R20291 (Stoke Mandeville) ribotype 027.
Potent enterotoxin.